missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum Copper spheres, alloy 2017, 9.5mm (0.37 in.) dia.

Brand:  Alfa Aesar™ 042034.KJ

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 5 pieces

 View more versions of this product

Product Code. 15486415

  • £33.71 / Pack of 5


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum Copper spheres, Alloy 2017
9.5mm (0.37 in.) dia.
MFCD00213972

Specifications

Al:Cu:Mn:Mg; 94.8:4:0.7:0.5 wt%
5 pieces
MFCD00213972
Aluminum Copper spheres, Alloy 2017
9.5mm (0.37 in.) dia.
Odorless
Videos
SDS