missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum Copper spheres, alloy 2017, 12.7mm (0.5 in.) dia.

Brand:  Alfa Aesar™ 042035.KG

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11315979

  • £22.58 / Each


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Specifications

Al:Cu:Mn:Mg; 94.8:4:0.7:0.5 wt%
1 piece
MFCD00213972
Aluminum Copper spheres, Alloy 2017
12.7mm (0.5 in.) dia.
Odorless
Videos
SDS