missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, Anhydrous, sublimed, ≥98%, Honeywell Fluka™

Brand:  Honeywell Fluka 11019-1KG

Code : NEW

Additional Details : CAS Number : 7446-70-0 Weight : 1.00000kg Transport : UN number : 1726 Chem class : 1a Pack group : II

Quantity 1kg

 View more versions of this product

Product Code. 15641000

  • £30.61 / 1kg


Estimated Shipment date
from Supplier 26-01-2018
Add to basket

Specifications

Chemical Identifiers

7446-70-0
≥98%
AlCl3
Anhydrous, Sublimed
1mmHg (100°C)
Aluminum Chloride
133.34g/mol
MFCD00003422
1726

Specifications

Aluminum Chloride
≥98%
133.34g/mol
1726
Anhydrous, Sublimed
7446-70-0
AlCl3
MFCD00003422
1mmHg (100°C)
1kg
Videos
SDS