missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)

Brand:  Alfa Aesar™ 010622.A1

Code : NEW

Additional Details : CAS Number : 7784-13-6 Weight : 1.00000kg Transport : UN number : 3260 Chem class : 8 Pack group : III

Quantity 1kg

 View more versions of this product

Product Code. 11360799

  • £574.90 / 1kg


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum chloride hexahydrate
Puratronic™
MFCD00149134
14,337
7784-13-6
241.43 (133.34 Anhydrous)
UN3260

Specifications

Aluminum chloride hexahydrate
Puratronic™
99.9995%
241.43 (133.34 Anhydrous)
UN3260
2.398g/mL
White
2.5
7784-13-6
Crystalline Aggregates
(metals basis)
MFCD00149134
14,337
Odorless
100°C (dec.)
1kg
Videos
SDS