missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Hexahydrate, 99%, Reagent Grade, Honeywell™

Catalog Number p-7120138
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7784-13-6
AlCl3H12O6
241.42
MFCD00149134
JGDITNMASUZKPW-UHFFFAOYSA-K
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
This item is not returnable. View return policy
7784-13-6
AlCl3H12O6
241.42
MFCD00149134
JGDITNMASUZKPW-UHFFFAOYSA-K
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Color White
pH 2.5 to 3.5 at 20°C
Quantity 100 g
UN Number NONH for all modes of transport
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 241.43g/mol
Percent Purity 99%
Grade ReagentPlus™
Packaging Poly Bottle
Appearance Crystals or Crystalline Chunk(s) or Chunk(s) or Solid
Vapor Pressure 1mmHg (100°C)
Density 2.4g/cm3
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'