missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Chloride, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Specifications
Melting Point | 190°C |
Color | White to Yellow |
Physical Form | Solid |
Quantity | 100 g |
Linear Formula | AlCl3 |
UN Number | UN1726 |
Formula Weight | 133.34g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Packaging | Glass Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'