missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum Beaker with Pourout Lip

Pour lip provides easy pouring of liquids into other containers.

Brand:  Alfa Aesar™ 039050.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15403695

  • £148.00 / Each


Estimated Shipment: 27-08-2021
Add to basket

Alert:

Not recommended for use at high temperatures


Description

Description

  • A highly impervious oxide film (approximately 5nm-thick) is resistant to corrosion by seawater, and other aqueous and chemical solutions
  • Aluminum offers chemical stability in the presence of most organic compounds .
Specifications

Specifications

With Pourout Lip
2000 mL
171 mm
Top: 137 mm; Bottom: 117 mm
Aluminum
Beaker
Videos
SDS Documents
Certificates