missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum acrylate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Aluminum acrylate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | soluble in acetone,benzene,carbon tetrachloride. Miscible with alcohol,ether,ethanol,ethyl ether,chloroform. |
Formula Weight | 243.17 |
Physical Form | Powder |
Chemical Name or Material | Aluminum acrylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'