missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum 2,4-pentanedionate, 99.995+% (metals basis), Thermo Scientific Chemicals
33.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 192°C to 194°C |
Density | 1.27 g/mL |
Boiling Point | 314°C to 316°C |
Quantity | 5 g |
UN Number | UN3467 |
Beilstein | 4157942 |
Formula Weight | 324.31 |
Percent Purity | ≥99.995% |
Physical Form | Powder |
Assay | (metals basis) |
Show More |
Safety and Handling
Product Identifier
- Aluminum 2,4-pentanedionate
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'