missing translation for 'onlineSavingsMsg'
Learn More

Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog Number p-7049031
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
13963-57-0
C15H21AlO6
324.31
MFCD00000013
KILURZWTCGSYRE-LNTINUHCSA-K
Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
This item is not returnable. View return policy
13963-57-0
C15H21AlO6
324.31
MFCD00000013
KILURZWTCGSYRE-LNTINUHCSA-K
Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Alumunium acetylacetonate may be used to prepare transparent superhydrophobic boehmite and silica films by sublimation, to deposit alumunium oxide films by chemical vapor deposition, to deposit alumunium oxide films by chemical vapor deposition, as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 192°C to 194°C
Density 1.27 g/mL
Boiling Point 314°C to 316°C
Quantity 100 g
UN Number UN3467
Beilstein 4157942
Solubility Information It is soluble in organic solvents but insoluble in water.
Formula Weight 324.31
Physical Form Powder
Chemical Name or Material Aluminum 2,4-pentanedionate
compliance-icons
Product Identifier
  • Aluminum 2,4-pentanedionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'