missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Alumunium acetylacetonate may be used to prepare transparent superhydrophobic boehmite and silica films by sublimation, to deposit alumunium oxide films by chemical vapor deposition, to deposit alumunium oxide films by chemical vapor deposition, as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 192°C to 194°C |
Density | 1.27 g/mL |
Boiling Point | 314°C to 316°C |
Quantity | 500 g |
UN Number | UN3467 |
Beilstein | 4157942 |
Solubility Information | It is soluble in organic solvents but insoluble in water. |
Formula Weight | 324.31 |
Physical Form | Powder |
Chemical Name or Material | Aluminum 2,4-pentanedionate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'