Learn More
Aluminon, ACS, Thermo Scientific Chemicals
Description
It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220°C to 225°C (decomposition) |
pH | 5.3 |
Odor | Odorless |
Quantity | 25 g |
Merck Index | 14,322 |
Solubility Information | Soluble in water(800g/L). |
Formula Weight | 473.44 |
Grade | ACS |
Physical Form | Powder |
Chemical Name or Material | Aluminon |
Safety and Handling
- Aluminon
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only