missing translation for 'onlineSavingsMsg'
Learn More

Aluminon, Thermo Scientific Chemicals

Catalog Number 10530994
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
569-58-4
C22H23N3O9
473.438
MFCD00040925
AIPNSHNRCQOTRI-UHFFFAOYSA-N
Aurintricarboxylic acid, triammonium salt
54729869
CHEBI:87398
triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]
This item is not returnable. View return policy
569-58-4
C22H23N3O9
473.438
MFCD00040925
AIPNSHNRCQOTRI-UHFFFAOYSA-N
Aurintricarboxylic acid, triammonium salt
54729869
CHEBI:87398
triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 220.0°C to 225.0°C
Beilstein 10, IV, 4161
Merck Index 15, 318
Solubility Information Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol, insoluble in petroleum ether
Infrared Spectrum Authentic
Formula Weight 473.44
Packaging Glass bottle
Suitability suitable for determination of aluminum
Color Red
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Aluminon
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'