Learn More
Aluminon, Thermo Scientific Chemicals
Use promo code "21687" to get your promotional price.
Description
It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types.
Solubility
Soluble in water(800g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 220°C to 225°C (decomposition) |
Merck Index | 14,322 |
Solubility Information | Soluble in water(800g/L). |
Formula Weight | 473.44 |
Odor | Odorless |
pH | 5.3 |
Quantity | 10 g |
Chemical Name or Material | Aluminon |
RUO – Research Use Only