missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum sulfate octadecahydrate, extra pure, Thermo Scientific Chemicals
178.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 86.0°C |
Color | Colorless to Yellow |
pH | 3 to 4 (2% soln. at 20°C) |
Physical Form | Liquid |
Quantity | 10 kg |
Linear Formula | Al2(SO4)3·18H2O |
Merck Index | 15,36 |
Solubility Information | Solubility in water: soluble. Other solubilities: practically insoluble in alcohol |
Formula Weight | 666.4 |
Percent Purity | 98.0 to 110.0% (Complexometry) |
Show More |
Safety and Handling
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'