missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog Number 10741535
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
10043-01-3
Al2O12S3
342.13
MFCD00003423
DIZPMCHEQGEION-UHFFFAOYSA-H
aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
24850
CHEBI:74768
dialuminum;trisulfate
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
10043-01-3
Al2O12S3
342.13
MFCD00003423
DIZPMCHEQGEION-UHFFFAOYSA-H
aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
24850
CHEBI:74768
dialuminum;trisulfate
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 770°C
Color White
Physical Form Chunks and/or Powder
Quantity 100 g
Assay Percent Range 15% min. (Al) Typical (Complexometry)
Linear Formula Al2(SO4)3
Merck Index 15, 360
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 342.13
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Aluminum sulfate, Extra Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'