missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Neutral, Brockmann Type I, for Chromatography, Camag 100-250 Mesh, Fisher Chemical

Catalog Number 10755321
Click to view available options
500g, Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy
Boiling Point 2980°C
Melting Point 2000°C
CAS 1344-28-1
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula Al2O3
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
Formula Weight 101.96g/mol
Packaging Plastic powder jar
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'