missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals

Catalog Number p-3734622
49.06 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]

Aluminum oxide, Al2O3, CAS Number-1344-28-1, abrasit, abrarex, abramax, abramant, alundum, Aluminum oxide, alpha-alumina, fasertonerde, Aluminum oxide, aloxite, 2.5kg, 2980.0 deg.C, White, 3.97g/mL, 215-691-6, 300ppm max.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for column chromatography
TRUSTED_SUSTAINABILITY
Boiling Point 2980.0°C
Melting Point 2050.0°C
Color White
pH 6.5 to 7.5 (5% aq. suspension, stirred)
Physical Form Fine Powder
Quantity 1 kg
Linear Formula Al2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15, 350
Solubility Information Solubility in water: insoluble
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade Chromatography
Packaging Plastic bottle
Density 3.97g/mL
Chemical Name or Material Aluminum oxide, Activated, Neutral, Brockmann I
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'