Learn More
Aluminium, technical, foil, ≈0.1mm thick, SLR, Fisher Chemical™
Brand: Fisher Chemical A/1570/53
Code : 21
Additional Details : CAS Number : 7429-90-5 Weight : 0.50000kg
Specifications
500g | |
Al | |
Plastic container | |
Solid Various Form | |
660°C |
7429-90-5 | |
134029 | |
26.97g/mol | |
Gray-Silver | |
2327°C |