missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acetylacetonate, 97%, Thermo Scientific Chemicals

Catalog Number p-3756523
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
13963-57-0
C15H21AlO6
324.31
MFCD00000013
KILURZWTCGSYRE-LNTINUHCSA-K
aluminum 2, 4-pentanedionate
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
This item is not returnable. View return policy
13963-57-0
C15H21AlO6
324.31
MFCD00000013
KILURZWTCGSYRE-LNTINUHCSA-K
aluminum 2, 4-pentanedionate
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 190.0°C to 195.0°C
Color White to Cream
Boiling Point 315.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Complexometry)
Packaging Glass bottle
Linear Formula (CH3COCH2COCH2)3Al
Quantity 5 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: 4.26 g/100g alcohol (20°C), 36.45 g/100g benzene (20°C), insoluble in petroleum ether
Formula Weight 324.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Aluminum acetylacetonate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium acetylacetonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'