missing translation for 'onlineSavingsMsg'
Learn More

Aluminium acetate, 90%, basic, ACROS Organics™

Brand:  Acros Organics 428190010

Code : 29

Additional Details : CAS Number : 142-03-0 Weight : 1.05000kg


Quantity 1kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10667020

  • £110.00 / 1kg


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

Aluminium acetate
142-03-0
100.0
C4H7AlO5
MFCD00008688
Basic
88.0
90%
162.08
Aluminium diacetate hydroxide, Aluminium hydroxide acetate

Specifications

Aluminium acetate
142-03-0
100.0
90%
162.08
Aluminium diacetate hydroxide, Aluminium hydroxide acetate
Solubility in water: slightly soluble.
Basic
88.0
90%
C4H7AlO5
MFCD00008688
Glass bottle
1kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aluminium acetate, basic

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS