missing translation for 'onlineSavingsMsg'
Learn More

Aluminium, 99%, powder, 200 mesh, ACROS Organics™

Brand:  Acros Organics 200930025

Code : 29

Additional Details : CAS Number : 7429-90-5 Weight : 2.55000kg Transport : UN number : 1396 Chem class : 4.3 Pack group : II


Product Code. 10763391

  • £68.90 / 2.50kg


EU Stock 8
Add to basket


Specifications

Specifications

Aluminium
200 mesh, 99%
300ppm max.
Finely Divided Powder
6000ppm max.
26.98
02,19; 04,9; 12,486; 16,337
Plastic bottle
5000ppm max.
Gray
2.5kg
7429-90-5
2.7
99%
Al
MFCD00134029
15, 319
75 Micron (>95% passes sieve)
800ppm max.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aluminium, powder

Signal Word
  • Danger

Hazard Category
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.
  • P402-Store in a dry place.

SDS