missing translation for 'onlineSavingsMsg'
Learn More

Aluminium, 99%, powder, 200 mesh, ACROS Organics™

Brand:  Acros Organics 200930250

Code : 29

Additional Details : CAS Number : 7429-90-5 Weight : 0.07500kg Transport : UN number : 1396 Chem class : 1 Pack group : II


Quantity 25g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10581641

  • £16.40 / 25g


EU Stock 11
Add to basket


Specifications

Chemical Identifiers

Aluminium
7429-90-5
Al
MFCD00134029
200 mesh, 99%
99%
26.98

Specifications

Aluminium
200 mesh, 99%
300ppm max.
Finely Divided Powder
6000ppm max.
26.98
02,19; 04,9; 12,486; 16,337
Plastic bottle
5000ppm max.
Gray
25g
7429-90-5
2.7
99%
Al
MFCD00134029
15, 319
75 Micron (>95% passes sieve)
800ppm max.
Videos
SDS