missing translation for 'onlineSavingsMsg'
Learn More

Aluminium, 99.97%, (trace metal basis), powder, ACROS Organics™

Brand:  Acros Organics 349040100

Code : 50

Additional Details : CAS Number : 7429-90-5 Weight : 0.06000kg Transport : UN number : 1396 Chem class : 1 Pack group : II


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10546841

  • £14.80 / 10g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Aluminium
7429-90-5
99.97%
26.98
99.97%
Trace Metal Basis
Al
MFCD00134029

Specifications

Aluminium
99.97%
2.7
Powder
Al
MFCD00134029
15, 319
0.03% max.
10g
7429-90-5
Trace Metal Basis
99.97%
26.98
02,19; 04,9; 12,486; 16,337
Glass bottle
Silvery-gray
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aluminium, powder

Signal Word
  • Danger

Hazard Category
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.
  • P402-Store in a dry place.

SDS