missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Alumina Wide Form Alsint Crucibles

Brand:  Haldenwanger™ 0B

Code : 02

Additional Details : Weight : 0.04000kg

 View more versions of this product

Product Code. 12041862

  • £31.02 / Each


Estimated Shipment
20-07-2018
Add to basket

Specifications

Specifications

25mL
37mm
Alumina
41mm
Alsint Crucible
Videos