missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Alumina Wide Form Alsint Crucibles

Brand:  Haldenwanger™ 00B

Code : 02

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 12031862

  • £21.73 / Each


Estimated Shipment
23-05-2018
Add to basket

Specifications

Specifications

10mL
24mm
Alumina
30mm
Alsint Crucible
Videos