missing translation for 'onlineSavingsMsg'
Learn More

Altanserin hydrochloride, Tocris Bioscience™

5-HT2A receptor antagonist

Brand:  Tocris (Bio-Techne) 180950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13450044

  • £557.13 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective 5-HT2A receptor antagonist (Ki values are 0.13, 4.55, 40, 62 and 1570 nM at 5-HT2A, α1, 5-HT2C, D2 and 5-HT1A respectively). Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

1135280-78-2
447.953
altanserin hydrochloride, unii-xul9717f5k, dsstox_cid_22592, dsstox_rid_80054, dsstox_gsid_42592, 4 1h-quinazolinone, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-, hydrochloride 1:1, 3-2-4-4-fluorobenzoyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorophenyl carbonyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone hydrochloride
3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-2-sulfanylidene-1H-quinazolin-4-one;hydrochloride
C22H23ClFN3O2S
JFPPLMAMMZZOEA-UHFFFAOYSA-N
24978536
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=S.Cl

Specifications

50mg
1135280-78-2
altanserin hydrochloride, unii-xul9717f5k, dsstox_cid_22592, dsstox_rid_80054, dsstox_gsid_42592, 4 1h-quinazolinone, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-, hydrochloride 1:1, 3-2-4-4-fluorobenzoyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorophenyl carbonyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone hydrochloride
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=S.Cl
447.953
447.95
Altanserin hydrochloride
C22H23ClFN3O2S
JFPPLMAMMZZOEA-UHFFFAOYSA-N
3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-2-sulfanylidene-1H-quinazolin-4-one;hydrochloride
24978536
>98%
Videos
Documents