Learn More
Alrestatin, Tocris Bioscience™
Aldose reductase inhibitor
Brand:  Tocris (Bio-Techne) 048510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13467723
  • £99.96 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Specific inhibitor of aldose reductase (IC50 = 148 μM). Attenuates glucose-induced angiotensin II production in rat vascular smooth muscle in vitro.
Specifications
Chemical Identifiers
51411-04-2
255.229
alrestatin, alrestatine, alrestatin inn, alrestatine inn-french, alrestatinum inn-latin, alrestatino inn-spanish, unii-515dhk15lg, spectrum_001449, tocris-0485, chembl63055
2-(1,3-dioxobenzo[de]isoquinolin-2-yl)acetic acid
C14H9NO4
GCUCIFQCGJIRNT-UHFFFAOYSA-N
2120
C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)CC(=O)O
Specifications
10mg
51411-04-2
alrestatin, alrestatine, alrestatin inn, alrestatine inn-french, alrestatinum inn-latin, alrestatino inn-spanish, unii-515dhk15lg, spectrum_001449, tocris-0485, chembl63055
C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)CC(=O)O
255.229
255.23
Alrestatin
C14H9NO4
GCUCIFQCGJIRNT-UHFFFAOYSA-N
2-(1,3-dioxobenzo[de]isoquinolin-2-yl)acetic acid
2120
>98%