missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ ALPI Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding mature form of human ALPI (Met 1- Asp 503), w/o pro peptide, fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 13225H02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15206285

  • £83.28 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

  • Formulation: Lyopihilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
79.5kd
5μg
Recombinant
Alkaline Phosphatase Intestinal
Assay Standard
Mammalian
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
ALPI
Other Proteins
ALPI Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.