Learn More
Ansell™ AlphaTec™ AquaDri™ Chemical Immersion Gloves
Catalog Number 15343442
Click to view available options
Size:
7
8
9
10
11
Two-layer construction keeps hands dryer for longer and with greater comfort and performance. Superior nitrile film provides advanced chemical protection against incidental exposure to bases, oils, fuels, some solvents, greases and animal fats.
  • Inner soft foam coating for moisture absorption
  • Nitrile film provides advanced chemical protection
  • Reversed-lozenge finish provides improved dry and wet grip
  • Nitrile compound for snag, puncture and abration protection
  • Foldable gutter cuff to prevent dripping on forearm
Back Material Nitrile
Cuff Material Nitrile
Product Type Immersion Gloves
Size 11
Style 58-335
Thickness (Metric) 0.3 mm
Glove Cuff Color Glass green
Glove Cuff Style Gauntlet
Liner Material Aquadri™ 2
Quantity 72 Pairs
Resistant To Chemical resistance
Series 58-335