missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Smooth Muscle Actin, clone: 1A4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9760-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15557646

  • £148.75 / 100µg


EU Stock 3
Add to basket

Description

Description

Description: The monoclonal antibody 1A4 recognizes human, mouse, and rat alpha-smooth muscle actin. Alpha-smooth muscle actin is a 42kDa protein and is a major component of the cytoskeletal structural network. This 1A4 antibody is specific for the alpha form of muscle actin only, which is expressed by smooth muscle cells of blood vessels, myofibroblasts, and myoepithelial cells. It is also expressed in the parenchyma and stroma of various tissues. Alpha-smooth muscle actin is useful in the identification of leiomyomas, leiomycosarcomas and pleomorphic adenomas and is used as a prognostic marker for basal cell carcinoma.Applications Reported: This 1A4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, immunoprecipitation, western blotting, immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry. (Fluorochrome conjugated 1A4 is recommended for use in intracellular flow cytometry.).Applications Tested: This 1A4 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low or high pH antigen retrieval and can be used at less than or equal to 1 μg/mL. This 1A4 antibody has been tested by immunocytochemistry of formaldehyde or methanol-fixed and permeabilized human cells and can be used at 1 μg/mL. This 1A4 antibody has been tested by western blot analysis of reduced rat and mouse intestine and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Alpha-Smooth Muscle Actin
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P62737, P62736, P62738
Actin, aortic smooth muscle, Alpha Actin 2, alpha-sma
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
HumanþMurineþRat
Flow CytometryþImmunocytochemistryþImmunohistochemistryþImmunohistochemistry (Frozen)þImmunoprecipitationþMicroscopyþWestern Blot
1A4
Unconjugated
ACTA2
Purified
Acta2
IgG2a, kappa
100μg
Store at 2-8°C.
Primary
59
Videos

For Research Use Only.