missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Naphtholphthalein, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Indicator in 0.1% or 0.04% alcohol solution.α-Naphtholphthalein is used as a pH indicator and operates in the pH ranges between 7.3 and 8.7. It is also used in the designing of optical carbon dioxide sensor based on fluorescence intensity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 238°C to 240°C |
Color | Brown |
Quantity | 5 g |
Merck Index | 14,6389 |
Solubility Information | Soluble in ethanol. Insoluble in water. |
Formula Weight | 418.45 |
Physical Form | Powder |
Chemical Name or Material | alpha-Naphtholphthalein |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'