missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamaldehyde, predominantly (E), 97%, Thermo Scientific Chemicals

Catalog Number p-7080757
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
101-39-3
C10H10O
146.19
MFCD00006976
VLUMOWNVWOXZAU-CLFYSBASSA-N
alpha-methylcinnamaldehyde, a-methylcinnamaldehyde, 2-propenal, 2-methyl-3-phenyl, 2-methyl-3-phenylprop-2-enal, e-2-methyl-3-phenylpropenal, 2-methyl-3-phenylacrylaldehyde, e-2-methyl-3-phenylacrylaldehyde, 2-methyl-3-phenyl-2-propenal, 2-methyl-3-phenylacrolein, alpha-methyl-trans-cinnamaldehyde
5372813
C\C(C=O)=C\C1=CC=CC=C1
This item is not returnable. View return policy
101-39-3
C10H10O
146.19
MFCD00006976
VLUMOWNVWOXZAU-CLFYSBASSA-N
alpha-methylcinnamaldehyde, a-methylcinnamaldehyde, 2-propenal, 2-methyl-3-phenyl, 2-methyl-3-phenylprop-2-enal, e-2-methyl-3-phenylpropenal, 2-methyl-3-phenylacrylaldehyde, e-2-methyl-3-phenylacrylaldehyde, 2-methyl-3-phenyl-2-propenal, 2-methyl-3-phenylacrolein, alpha-methyl-trans-cinnamaldehyde
5372813
C\C(C=O)=C\C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.05
Boiling Point 138°C to 140°C (14 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.605
Quantity 100 g
Beilstein 507514
Sensitivity Air Sensitive
IUPAC Name (E)-2-methyl-3-phenylprop-2-enal
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material alpha-Methylcinnamaldehyde, predominantly ( E)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'