missing translation for 'onlineSavingsMsg'
Learn More

alpha GalCer:CD1d Complex, Functional Grade, clone: L363, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-2019-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298587

  • £56.31 / 25µg


Estimated Shipment
02-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation.The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes.Applications Reported: This L363 antibody has been reported for use in functional assays.Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylcermide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

alpha GalCer:CD1d complex
L363
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Functional Assay
1mg/mL
aqueous buffer, no sodium azide
alpha GalCer-CD1d complex
IgG2a
RUO
Primary
Murine
Videos