missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-D-Lactose, monohydrate, ACS reagent, ACROS Organics™
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic bucket
Specifications
Color | White |
Ignition Residue | 0.03% max. |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Quantity | 500 g |
Beilstein | 31, 408 |
Merck Index | 14, 5343 |
Specific Rotation Condition | + 54.40 (20.00°C c=10,H2O,NH3) |
Solubility Information | 0.005% max. Insoluble Matter |
Formula Weight | 360.3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'