missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Chloralose, 98+%, beta anomer ca 20%, Thermo Scientific Chemicals

Catalog Number p-7122665
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
15879-93-3
C8H11Cl3O6
309.52
MFCD00005542
OJYGBLRPYBAHRT-OPKHMCHVSA-N
a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol
7057995
OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl
This item is not returnable. View return policy
15879-93-3
C8H11Cl3O6
309.52
MFCD00005542
OJYGBLRPYBAHRT-OPKHMCHVSA-N
a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol
7057995
OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canadensis) and blue geese (Chen caerulescens) were caught with alpha-chloralose, an oral hypnotic administered on bait. Wild turkeys (Meleagris gallopavo) were captured with alpha-chloralose, an oral anesthetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canaden
TRUSTED_SUSTAINABILITY
Melting Point 180°C to 184°C
Quantity 25 g
Beilstein 85418
Merck Index 14,2072
Solubility Information Slightly soluble in water.
IUPAC Name 1-[(2R,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
Formula Weight 309.53
Percent Purity ≥98%
Chemical Name or Material alpha-Chloralose, beta anomer ≈ 15%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'