Learn More
Alpha Catenin Mouse anti-Human, Clone: 1G5, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-9120-82
Code : NEW
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 15547586
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
The monoclonal antibody 1G5 recognizes human alpha catenin. Alpha catenin plays a role in cell-cell adhesion through its function in the binding and bundling of actin. The N-terminus of alpha catenin binds beta catenin and the C-terminus binds actin. Monomeric alpha catenin acts as a physical linker between the cadherin-beta catenin complex and the actin cytoskeleton and is stabilized by dimeric alpha catenin found in the cytoplasm. Alpha catenin also binds other actin-related proteins, including vinculin, alpha-actinin, ZO-1, afadin, formin-1, and Rho. Alpha catenin is absent in some tumor cell lines and reduced in some human carcinomas.
Specifications
Specifications
Alpha Catenin
1G5
Unconjugated
Purified
IgG1, kappa
RUO
Primary
Human
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C.