missing translation for 'onlineSavingsMsg'
Learn More

α-Bromocinnamaldehyde, 98%, Thermo Scientific™

Catalog Number 11964801
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
5443-49-2
WQRWNOKNRHCLHV-TWGQIWQCSA-N
alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo
5369403
(Z)-2-bromo-3-phenylprop-2-enal
C1=CC=C(C=C1)C=C(C=O)Br
This item is not returnable. View return policy
5443-49-2
WQRWNOKNRHCLHV-TWGQIWQCSA-N
alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo
5369403
(Z)-2-bromo-3-phenylprop-2-enal
C1=CC=C(C=C1)C=C(C=O)Br
Melting Point 68.0°C to 72.0°C
Color Yellow to Ochre
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H7BrO
Linear Formula C6H5CH=C(Br)CHO
Quantity 1 kg
Beilstein 07,358
Molecular Weight (g/mol) 211.06
Formula Weight 211.06
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Bromocinnamaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromocinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'