missing translation for 'onlineSavingsMsg'
Learn More

alpha beta TCR Mouse anti-Human, PerCP-eFluor 710, Clone: IP26, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9986-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15599476

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR.The alpha beta TCR is expressed by the majority of peripheral T cells.
Specifications

Specifications

alpha beta TCR
IP26
PerCP-eFluor 710
Conjugated
IgG1, kappa
RUO
Primary
Human
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos
Documents

For Research Use Only.