missing translation for 'onlineSavingsMsg'
Learn More

TCR alpha/beta, clone: IP26, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9986-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238317

  • £31.01 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR.The alpha beta TCR is expressed by the majority of peripheral T cells.Applications Reported: The IP26 antibody has been reported for use in flow cytometric analysis.Applications Tested: The IP26 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

alpha beta TCR
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01848, P04435
Tcrb, Tcra, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
IP26
Unconjugated
TCRB
Purified
TCRA, TCRB
IgG1, kappa
25μg
Store at 2-8°C.
Primary
6955, 6957
Videos

For Research Use Only.