missing translation for 'onlineSavingsMsg'
Learn More

α,α-Diphenyl-4-piperidinomethanol hydrochloride, 98%, Acros Organics™

Brand:  Acros Organics 124981000

Code : 29

Additional Details : CAS Number : 1798-50-1 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10396520

  • £48.30 / 100g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

α,α-Diphenyl-4-piperidinomethanol hydrochloride
1798-50-1
100.0
98%
303.82
98%
97.5
98%
C18H21NO·HCl
MFCD00039039

Specifications

α,α-Diphenyl-4-piperidinomethanol hydrochloride
270.0° to 281.0°C
98%
97.5
Authentic
Glass bottle
98%
C18H21NO·HCl
MFCD00039039
White to off-white
100g
1798-50-1
100.0
10% max.
Crystalline Powder
98%
303.82
13, 895
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • a,a-Diphenyl-4-piperidinomethanol hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS