missing translation for 'onlineSavingsMsg'
Learn More

α,α-Diphenyl-4-piperidinomethanol hydrochloride, 98%, Acros Organics™

Brand:  Acros Organics 124980250

Code : 29

Additional Details : CAS Number : 1798-50-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10336480

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

α,α-Diphenyl-4-piperidinomethanol hydrochloride
1798-50-1
100.0
98%
303.82
98%
97.5
98%
C18H21NO·HCl
MFCD00039039

Specifications

α,α-Diphenyl-4-piperidinomethanol hydrochloride
270.0° to 281.0°C
98%
97.5
Authentic
Glass bottle
98%
C18H21NO·HCl
MFCD00039039
White to off-white
25g
1798-50-1
100.0
10% max.
Crystalline Powder
98%
303.82
13, 895
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • a,a-Diphenyl-4-piperidinomethanol hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS