missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trifluorotoluene, 99+%, Thermo Scientific Chemicals

Catalog Number p-204946
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Plastic bottle
98-08-8
C7H5F3
146.11
MFCD00000372
GETTZEONDQJALK-UHFFFAOYSA-N
benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
7368
CHEBI:36810
trifluoromethylbenzene
C1=CC=C(C=C1)C(F)(F)F
98-08-8
C7H5F3
146.11
MFCD00000372
GETTZEONDQJALK-UHFFFAOYSA-N
benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
7368
CHEBI:36810
trifluoromethylbenzene
C1=CC=C(C=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -29°C
Color Colorless
Density 1.1900g/mL
Boiling Point 102°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CF3
Refractive Index 1.4130 to 1.415
Quantity 50 mL
Beilstein 05, 290
Merck Index 15, 1113
Specific Gravity 1.19
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in many organic solvents
Formula Weight 146.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluorotoluene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chromosulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1B
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, Potassium dichromate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'