missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha’,alpha’-Tetramethyl-5-bromomethyl-1,3-benzenediacetonitrile, TRC

Product Code. 30410874
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410874 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410874 Supplier Toronto Research Chemicals Supplier No. TRCT291490100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha,alpha,alpha',alpha'-Tetramethyl-5-bromomethyl-1,3-benzenediacetonitrile
CAS 120511-84-4
Molecular Formula C15 H17 Br N2
Quantity 100 mg
Synonym 5-(Bromomethyl)-α1,α1,α3,α3-tetramethyl-1,3-benzenediacetonitrile, 1,3-Benzenediacetonitrile','5-(bromomethyl)-α,α,α',α'-tetramethyl- (9CI), 5-(Bromomethyl)-α1,α1,α3,α3-tetramethyl-1,3-benzenediacetonitrile, 3,5-Bis(1-cyano-1-methylethyl)benzyl bromide, 3,5-Bis(2-cyanoprop-2-yl)benzyl bromide, 2,2'-[5-(Bromomethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile)
InChI Formula InChI=1S/C15H17BrN2/c1-14(2,9-17)12-5-11(8-16)6-13(7-12)15(3,4)10-18/h5-7H,8H2,1-4H3
SMILES CC(C)(C#N)c1cc(CBr)cc(c1)C(C)(C)C#N
IUPAC Name 2-[3-(bromomethyl)-5-(2-cyanopropan-2-yl)phenyl]-2-methylpropanenitrile
Molecular Weight (g/mol) 305.21
Formula Weight 304.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.