missing translation for 'onlineSavingsMsg'
Learn More

alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane, 99%, Thermo Scientific Chemicals

Catalog Number p-7033877
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
167482-98-6
C19H22N2
278.399
MFCD03454220
SLPZWMKFAWAFSE-UHFFFAOYSA-N
5-4-tert-butylphenyl dipyrromethane, alpha-4-tert-butylphenyl di 2-pyrrolyl methane, e'-4-tert-butylphenyl di 2-pyrrolyl methane, 2-4-tert-butylphenyl 1h-pyrrol-2-yl methyl-1h-pyrrole, a-4-tert-butylphenyl di 2-pyrrolyl methane, 2,2'-4-tert-butylphenyl methylenebispyrrole, 2,2'-4-tert-butylbenzylidene bis 1h-pyrrole, 2,2'-4-tert-butylphenyl methylene di 1h-pyrrole, 2-4-tert-butylphenyl-1h-pyrrol-2-yl methyl-1h-pyrrole, 1h-pyrrole, 2,2'-4-1,1-dimethylethyl phenyl methylene bis
4245355
2-[(4-tert-butylphenyl)-(1H-pyrrol-2-yl)methyl]-1H-pyrrole
CC(C)(C)C1=CC=C(C=C1)C(C2=CC=CN2)C3=CC=CN3
This item is not returnable. View return policy
167482-98-6
C19H22N2
278.399
MFCD03454220
SLPZWMKFAWAFSE-UHFFFAOYSA-N
5-4-tert-butylphenyl dipyrromethane, alpha-4-tert-butylphenyl di 2-pyrrolyl methane, e'-4-tert-butylphenyl di 2-pyrrolyl methane, 2-4-tert-butylphenyl 1h-pyrrol-2-yl methyl-1h-pyrrole, a-4-tert-butylphenyl di 2-pyrrolyl methane, 2,2'-4-tert-butylphenyl methylenebispyrrole, 2,2'-4-tert-butylbenzylidene bis 1h-pyrrole, 2,2'-4-tert-butylphenyl methylene di 1h-pyrrole, 2-4-tert-butylphenyl-1h-pyrrol-2-yl methyl-1h-pyrrole, 1h-pyrrole, 2,2'-4-1,1-dimethylethyl phenyl methylene bis
4245355
2-[(4-tert-butylphenyl)-(1H-pyrrol-2-yl)methyl]-1H-pyrrole
CC(C)(C)C1=CC=C(C=C1)C(C2=CC=CN2)C3=CC=CN3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 158°C to 162°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 278.39
Percent Purity 99%
Chemical Name or Material alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'