missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium Bromide 99%, ACROS Organics™

Brand:  Acros Organics 103040050

Code : 29

Additional Details : CAS Number : 1560-54-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10456422

  • £9.40 / 5g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

Allyltriphenylphosphonium bromide
1560-54-9
100.0
99%
H2C=CHCH2P(C6H5)3Br
MFCD00011808
99%
98.5
99%
C21H20BrP
383.26

Specifications

Allyltriphenylphosphonium bromide
1560-54-9
100.0
Glass bottle
99%
C21H20BrP
383.26
White
5g
99%
98.5
Authentic
Powder
99%
H2C=CHCH2P(C6H5)3Br
MFCD00011808
222.0° to 225.0°C
Videos
SDS