missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethoxysilane, 97+%, Thermo Scientific Chemicals

Catalog Number p-3586386
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2551-83-9
C6H14O3Si
162.26
MFCD00053865
LFRDHGNFBLIJIY-UHFFFAOYSA-N
allyltrimethoxysilane, silane, trimethoxy-2-propenyl, silane, trimethoxy-2-propen-1-yl, allyl trimethoxy silane, silane, allyltrimethoxy, allytrimethoxysilane, acmc-209gkn, trimethoxy prop-2-enyl silane, silane,trimethoxy-2-propen-1-yl
75698
trimethoxy(prop-2-enyl)silane
CO[Si](CC=C)(OC)OC
This item is not returnable. View return policy
2551-83-9
C6H14O3Si
162.26
MFCD00053865
LFRDHGNFBLIJIY-UHFFFAOYSA-N
allyltrimethoxysilane, silane, trimethoxy-2-propenyl, silane, trimethoxy-2-propen-1-yl, allyl trimethoxy silane, silane, allyltrimethoxy, allytrimethoxysilane, acmc-209gkn, trimethoxy prop-2-enyl silane, silane,trimethoxy-2-propen-1-yl
75698
trimethoxy(prop-2-enyl)silane
CO[Si](CC=C)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 146°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2Si(OCH3)3
Refractive Index 1.4040 to 1.406
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: reacts
Formula Weight 162.26
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Allyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'