missing translation for 'onlineSavingsMsg'
Learn More

Allyltrichlorosilane, 95%, Thermo Scientific™

Catalog Number p-52299
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
107-37-9
C3H5Cl3Si
175.52
MFCD00000486
HKFSBKQQYCMCKO-UHFFFAOYSA-N
allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl
7867
trichloro(prop-2-enyl)silane
C=CC[Si](Cl)(Cl)Cl
This item is not returnable. View return policy
107-37-9
C3H5Cl3Si
175.52
MFCD00000486
HKFSBKQQYCMCKO-UHFFFAOYSA-N
allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl
7867
trichloro(prop-2-enyl)silane
C=CC[Si](Cl)(Cl)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 117.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2CCHCH2SiCl3
Refractive Index 1.4430 to 1.4470
Quantity 5 g
Specific Gravity 1.2
Solubility Information Solubility in water: react violently with water
Formula Weight 175.52
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Allyltrichlorosilane, 95%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'