missing translation for 'onlineSavingsMsg'
Learn More

Allylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-52208
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
107-11-9
C3H7N
57.10
MFCD00008199
VVJKKWFAADXIJK-UHFFFAOYSA-N
allylamine, 2-propen-1-amine, monoallylamine, 3-aminopropylene, 3-aminopropene, allyl amine, 2-propenamine, 3-amino-1-propene, 2-propenylamine, polyallylamine
7853
prop-2-en-1-amine
NCC=C
This item is not returnable. View return policy
107-11-9
C3H7N
57.10
MFCD00008199
VVJKKWFAADXIJK-UHFFFAOYSA-N
allylamine, 2-propen-1-amine, monoallylamine, 3-aminopropylene, 3-aminopropene, allyl amine, 2-propenamine, 3-amino-1-propene, 2-propenylamine, polyallylamine
7853
prop-2-en-1-amine
NCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -88°C
Color White
Density 0.7630g/mL
Boiling Point 53°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2NH2
Refractive Index 1.4205 to 1.4225
Quantity 50 mL
Beilstein 04,205
Merck Index 15,278
Specific Gravity 0.763
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether
Viscosity 0.44 mPa.s (25°C)
Formula Weight 57.09
Percent Purity 98%
Grade Extra Pure
Physical Form Crystalline or Granular Powder
Chemical Name or Material Allylamine, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'