missing translation for 'onlineSavingsMsg'
Learn More

Allyl trifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7081641
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
383-67-5
C5H5F3O2
154.088
MFCD00013567
XIVPVSIDXBTZLM-UHFFFAOYSA-N
allyl trifluoroacetate, allyl trifluoracetate, trifluoroacetic acid allyl ester, prop-2-en-1-yl 2,2,2-trifluoroacetate, allyltrifluoroacetate, pubchem12548, acmc-209izu, allyl alcohol, trifluoroacetate, acetic acid, trifluoro-, allyl ester
67845
prop-2-enyl 2,2,2-trifluoroacetate
C=CCOC(=O)C(F)(F)F
This item is not returnable. View return policy
383-67-5
C5H5F3O2
154.088
MFCD00013567
XIVPVSIDXBTZLM-UHFFFAOYSA-N
allyl trifluoroacetate, allyl trifluoracetate, trifluoroacetic acid allyl ester, prop-2-en-1-yl 2,2,2-trifluoroacetate, allyltrifluoroacetate, pubchem12548, acmc-209izu, allyl alcohol, trifluoroacetate, acetic acid, trifluoro-, allyl ester
67845
prop-2-enyl 2,2,2-trifluoroacetate
C=CCOC(=O)C(F)(F)F

Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Density 1.183
Boiling Point 73°C to 75°C
Flash Point −1°C (30°F)
Refractive Index 1.336
Quantity 10 g
UN Number UN2924
Beilstein 1766312
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 154.09
Percent Purity 97%
Chemical Name or Material Allyl trifluoroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'